Diethyl benzylmalonate, 97%
Diethyl benzylmalonate, 97%
Diethyl benzylmalonate, 97%
Thermo Scientific Chemicals

Diethyl benzylmalonate, 97%

CAS: 607-81-8 | C14H18O4 | 250.294 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number B22890.14
also known as B22890-14
Price (EUR)
34,50
Each
Quantity:
25 g
Request bulk or custom format
Price (EUR)
34,50
Each
Chemical Identifiers
CAS607-81-8
IUPAC Name1,3-diethyl 2-benzylpropanedioate
Molecular FormulaC14H18O4
InChI KeyICZLTZWATFXDLP-UHFFFAOYSA-N
SMILESCCOC(=O)C(CC1=CC=CC=C1)C(=O)OCC
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.4840-1.4890 @ 20?C
FormLiquid
Appearance (Color)Clear colorless
Assay (GC)≥96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only