2-Phenylbutyryl chloride, 98%, Thermo Scientific Chemicals
2-Phenylbutyryl chloride, 98%, Thermo Scientific Chemicals
2-Phenylbutyryl chloride, 98%, Thermo Scientific Chemicals
2-Phenylbutyryl chloride, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Phenylbutyryl chloride, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number B22997.14
also known as B22997-14
Price (EUR)
-
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS36854-57-6
IUPAC Name2-phenylbutanoyl chloride
Molecular FormulaC10H11ClO
InChI KeyQGXMHCMPIAYMGT-UHFFFAOYNA-N
SMILESCCC(C(Cl)=O)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.5150-1.5190 @ 20?C
Appearance (Color)Clear colorless
Assay (GC)≥97.5%
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only