(R)-(-)-2,3-O-Isopropylideneglycerol, 96%, Thermo Scientific Chemicals
(R)-(-)-2,3-O-Isopropylideneglycerol, 96%, Thermo Scientific Chemicals
(R)-(-)-2,3-O-Isopropylideneglycerol, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

(R)-(-)-2,3-O-Isopropylideneglycerol, 96%, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
B23037.06
also known as B23037-06
5 g
Catalog number B23037.06
also known as B23037-06
Price (EUR)
107,00
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
107,00
Each
Chemical Identifiers
CAS14347-78-5
IUPAC Name(2,2-dimethyl-1,3-dioxolan-4-yl)methanol
Molecular FormulaC6H12O3
InChI KeyRNVYQYLELCKWAN-UHFFFAOYNA-N
SMILESCC1(C)OCC(CO)O1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
FormLiquid
Assay (GC)≥95.0%
Optical Rotation-11.5 ± 0.3? (C=5 in methanol)
Refractive Index1.4320-1.4380 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only