5-Methylthiophene-2-boronic acid, 98%
5-Methylthiophene-2-boronic acid, 98%
5-Methylthiophene-2-boronic acid, 98%
Thermo Scientific Chemicals

5-Methylthiophene-2-boronic acid, 98%

CAS: 162607-20-7 | C5H7BO2S | 141.98 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number B23138.06
also known as B23138-06
Price (EUR)
152,00
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
152,00
Each
Chemical Identifiers
CAS162607-20-7
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)≥97.5%
Proton NMRConforms to structure
FormCrystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)≥97.5%
Appearance (Color)White to cream
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only