2-(Methylamino)ethanol, 99%
2-(Methylamino)ethanol, 99%
2-(Methylamino)ethanol, 99%
Thermo Scientific Chemicals

2-(Methylamino)ethanol, 99%

CAS: 109-83-1 | C3H9NO | 75.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
Catalog number B23180.AE
also known as B23180-AE
Price (EUR)
32,80
Each
Quantity:
100 mL
Request bulk or custom format
Price (EUR)
32,80
Each
Chemical Identifiers
CAS109-83-1
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.4380-1.4410 @ 20?C
Assay (GC)≥98.5%
Appearance (Color)Clear colorless to pale yellow
FormLiquid
2-(Methylamino)ethanol is used as an intermediate in synthetic chemistry. It finds application in various fields such as textile lubricants, polishes, detergents and in personal care products. It is also involved in electrostatic automotive coatings and in acid gas scrubbing. Further, it serves as an intermediate in polymers and pharmaceuticals. In addition to this, it is used as a solvent in the natural gas-processing industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(Methylamino)ethanol is used as an intermediate in synthetic chemistry. It finds application in various fields such as textile lubricants, polishes, detergents and in personal care products. It is also involved in electrostatic automotive coatings and in acid gas scrubbing. Further, it serves as an intermediate in polymers and pharmaceuticals. In addition to this, it is used as a solvent in the natural gas-processing industry.

Solubility
Miscible with water, alcohol and ether.

Notes
Incompatible with oxidizing agents, acids, copper, copper alloys, zinc and galvanized iron.
RUO – Research Use Only

General References:

  1. Khusnutdinova, J. R.; Garg, J. A.; Milstein, D. Combining Low-Pressure CO2 Capture and Hydrogenation To Form Methanol. ACS Catal. 2015, 5 (4), 2416-2422.
  2. Fytianos, G.; Callot, R.; Svendsen, H. F.; Knuutila, H. K. Quantitative determination of amines used in post-combustion CO2 capture process by ion chromatography. Int. J. Greenhouse Gas Control 2015, 42, 372-378.