(+/-)-2-Phenylbutyric acid, 98%, Thermo Scientific Chemicals
(+/-)-2-Phenylbutyric acid, 98%, Thermo Scientific Chemicals
(+/-)-2-Phenylbutyric acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

(+/-)-2-Phenylbutyric acid, 98%, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
B23222.22
also known as B23222-22
100 g
Catalog number B23222.22
also known as B23222-22
Price (EUR)
49,65
Online exclusive
55,40
Save 5,75 (10%)
Each
Quantity:
100 g
Request bulk or custom format
Price (EUR)
49,65
Online exclusive
55,40
Save 5,75 (10%)
Each
Chemical Identifiers
CAS90-27-7
IUPAC Name2-phenylbutanoic acid
Molecular FormulaC10H12O2
InChI KeyOFJWFSNDPCAWDK-UHFFFAOYNA-N
SMILESCCC(C(O)=O)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
FormCrystals or powder or crystalline powder
Assay (Silylated GC)≥97.5%
Melting Point (clear melt)39.0-45.0?C
Optical Rotation0.0 ± 0.2? (C = 1 in MeOH)
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only