N-Methyl-1,3-propanediamine, 99%
N-Methyl-1,3-propanediamine, 99%
N-Methyl-1,3-propanediamine, 99%
N-Methyl-1,3-propanediamine, 99%
Thermo Scientific Chemicals

N-Methyl-1,3-propanediamine, 99%

CAS: 6291-84-5 | C4H12N2 | 88.154 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number B23319.36
also known as B23319-36
Price (EUR)
133,00
Each
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (EUR)
133,00
Each
Add to cart
Chemical Identifiers
CAS6291-84-5
IUPAC Name(3-aminopropyl)(methyl)amine
Molecular FormulaC4H12N2
InChI KeyQHJABUZHRJTCAR-UHFFFAOYSA-N
SMILESCNCCCN
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Appearance (Color)Clear colorless
Refractive Index1.4450-1.4480 @ 20?C
Assay (GC)≥98.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only