2,2'-Dimethoxybiphenyl, 97%
2,2'-Dimethoxybiphenyl, 97%
2,2'-Dimethoxybiphenyl, 97%
Thermo Scientific Chemicals

2,2'-Dimethoxybiphenyl, 97%

CAS: 4877-93-4 | C14H14O2 | 214.26 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number B23411.14
also known as B23411-14
Price (EUR)
137,00
Each
Quantity:
25 g
Request bulk or custom format
Price (EUR)
137,00
Each
Chemical Identifiers
CAS4877-93-4
IUPAC Name2,2'-dimethoxy-1,1'-biphenyl
Molecular FormulaC14H14O2
InChI KeyVGMKUVCDINAAFC-UHFFFAOYSA-N
SMILESCOC1=CC=CC=C1C1=CC=CC=C1OC
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Melting Point (clear melt)151.0-160.0?C
Assay (GC)≥96.0%
Appearance (Color)White to pale cream
2,2-Dimethoxy-1,1-biphenyl is used in the synthesis of biphenyl scaffolds in the preparation of biphenyl-tetrathiafulvalene derivatives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,2-Dimethoxy-1,1-biphenyl is used in the synthesis of biphenyl scaffolds in the preparation of biphenyl-tetrathiafulvalene derivatives.

Solubility
Soluble in chloroform, methanol.

Notes
Store in a cool, dry conditions, in well sealed conditions. Store away from oxidizing agents.
RUO – Research Use Only

General References:

  1. Noriyuki Yonezawa; Shiro Miyata; Toshiaki Nakamura; Shinichiro Mori; Yuriko Ueha; Ryoichi Katakai. Synthesis of wholly aromatic polyketones using 2,2'-dimethoxybiphenyl as the acyl-acceptant monomer. Macromolecules. 1993, 26 (19), 5262-5263.
  2. N. P. Buu-Hoë; L. Petit; D. C. Thang. Orientation in Some Friedel—Crafts Acylations of 2,2'-Dimethoxybiphenyl, and the Cyclization of the Reaction Products. J. Org. Chem. 1964, 29 (3), 762-763.