1,3-Benzenedimethanol, 98%
1,3-Benzenedimethanol, 98%
1,3-Benzenedimethanol, 98%
Thermo Scientific Chemicals

1,3-Benzenedimethanol, 98%

CAS: 626-18-6 | C8H10O2 | 138.166 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number B23417.14
also known as B23417-14
Price (EUR)
331,00
Each
Quantity:
25 g
Request bulk or custom format
Price (EUR)
331,00
Each
Chemical Identifiers
CAS626-18-6
IUPAC Name[3-(hydroxymethyl)phenyl]methanol
Molecular FormulaC8H10O2
InChI KeyYWMLORGQOFONNT-UHFFFAOYSA-N
SMILESOCC1=CC(CO)=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream
Assay (GC)≥97.5%
FormCrystalline powder and/or chunks
Melting Point (clear melt)53-60?C
1,3-Benzenedimethanol was used in the synthesis of a series of cross-linked poly(orthocarbonate)s. It was also used in the synthesis of mixed- tethered system, required for the preparation of e-edge-[60]fullerenylmethanodihydropyrrole adduct.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,3-Benzenedimethanol was used in the synthesis of a series of cross-linked poly(orthocarbonate)s. It was also used in the synthesis of mixed- tethered system, required for the preparation of e-edge-[60]fullerenylmethanodihydropyrrole adduct.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agent.
RUO – Research Use Only

General References:

  1. Sonmez HB and Wudl F. Cross-linked poly (orthocarbonate) s as organic solvent sorbents. Macromolecules. 2005, 38 (5), 1623-1626.
  2. Leila Chaker; Graham E. Ball; James R. Williams; Glenn A. Burley; Bill C. Hawkins; Paul A. Keller and Stephen G. Pyne. Regioselective Synthesis of Novel e-Edge-[60]fullerenylmethanodihydropyrroles and 1,2-Dihydromethano[60]fullerenes. European J. Org. Chem. 2005, 24 (3), 5158-5162.