Ethyl octanoate, 99%
Ethyl octanoate, 99%
Ethyl octanoate, 99%
Thermo Scientific Chemicals

Ethyl octanoate, 99%

CAS: 106-32-1 | C10H20O2 | 172.27 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number B23450.22
also known as B23450-22
Price (EUR)
25,50
Each
Quantity:
100 g
Request bulk or custom format
Price (EUR)
25,50
Each
Chemical Identifiers
CAS106-32-1
IUPAC Nameethyl octanoate
Molecular FormulaC10H20O2
InChI KeyYYZUSRORWSJGET-UHFFFAOYSA-N
SMILESCCCCCCCC(=O)OCC
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Appearance (Color)Clear colorless
Assay (GC)≥98.5 to ≤101.5%
Refractive Index1.4155-1.4185 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only