4-Amino-3-nitrobenzonitrile, 98%
4-Amino-3-nitrobenzonitrile, 98%
4-Amino-3-nitrobenzonitrile, 98%
Thermo Scientific Chemicals

4-Amino-3-nitrobenzonitrile, 98%

CAS: 6393-40-4 | C7H5N3O2 | 163.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number B23463.03
also known as B23463-03
Price (EUR)
28,80
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
28,80
Each
Chemical Identifiers
CAS6393-40-4
IUPAC Name4-amino-3-nitrobenzonitrile
Molecular FormulaC7H5N3O2
InChI KeyJAHADAZIDZMHOP-UHFFFAOYSA-N
SMILESNC1=CC=C(C=C1[N+]([O-])=O)C#N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow
Melting Point (clear melt)156.0-163.0?C
Assay (GC)≥97.5%
FormPowder
4-Amino-3-nitrobenzonitrile is used in gamma irradiations effect on release of this compound from polyanhydride matrixes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Amino-3-nitrobenzonitrile is used in gamma irradiations effect on release of this compound from polyanhydride matrixes.

Solubility
Soluble in ethanol and benzene.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Batista, R. M. F.; Ferreira, R. C. M.; Raposo, M. M. M.; Costa, S. P. G. Novel optical chemosensors for anions and cations based on an amino acid core functionalised with benzimidazoles. Tetrahedron 2012, 68 (36), 7322-7330.
  2. Stolić, I.; Bajić, M.; Matković-Čalogović, D. Effect of Hydrogen Bonding on the Pyramidalization of the Amino Group: Structure of 3,4-diaminobenzamidinium Chloride. Croat. Chem. Acta 2013, 86 (2), 193-199.