1,2-Dimethylimidazole, 98%
1,2-Dimethylimidazole, 98%
1,2-Dimethylimidazole, 98%
Thermo Scientific Chemicals

1,2-Dimethylimidazole, 98%

CAS: 1739-84-0 | C5H8N2 | 96.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number B23544.22
also known as B23544-22
Price (EUR)
43,30
Each
Quantity:
100 g
Request bulk or custom format
Price (EUR)
43,30
Each
Chemical Identifiers
CAS1739-84-0
IUPAC Name1,2-dimethyl-1H-imidazole
Molecular FormulaC5H8N2
InChI KeyGIWQSPITLQVMSG-UHFFFAOYSA-N
SMILESCN1C=CN=C1C
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥97.5%
Appearance (Color)Colorless to white to pale yellow
Melting Point (clear melt)27.0-40.0?C
FormCrystals or powder or crystalline powder or fused solid or chunks
Identification (FTIR)Conforms
1,2-Dimethylimidazole is used in the synthesis of 1,2-dimethyl-3-n-butylimidazoliumchloride and 1,2-dimethyl-3-n-propylimidazolium chloride. It also can be used in the synthesis of 1-(2-methoxyethyl)-2,3-dimethylimidazolium chloride and hexafluorophosphate salts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,2-Dimethylimidazole is used in the synthesis of 1,2-dimethyl-3-n-butylimidazoliumchloride and 1,2-dimethyl-3-n-propylimidazolium chloride. It also can be used in the synthesis of 1-(2-methoxyethyl)-2,3-dimethylimidazolium chloride and hexafluorophosphate salts.

Solubility
Soluble in water.

Notes
Incompatible materials are oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
RUO – Research Use Only

General References:

  1. Wesley A Henderson; Victor G Young; Douglas M Fox; Hugh C De Long; Paul C Trulove. Alkyl vs. alkoxy chains on ionic liquid cations. Chemical Communications. 2006, (35), 3708-3710.
  2. A.Latif Abuhijleh. Synthesis and molecular structure of monomeric copper(II) acetates with 2-methylimidazole and 1,2-dimethylimidazole. Inorganica Chimica Acta. 1991, 190, 11-17.
  3. Buffer: Biochemistry, 12, 5127 (1973).