meso-Hydrobenzoin, 95%
meso-Hydrobenzoin, 95%
meso-Hydrobenzoin, 95%
Thermo Scientific Chemicals

meso-Hydrobenzoin, 95%

CAS: 579-43-1 | C14H14O2 | 214.26 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number B23946.06
also known as B23946-06
Price (EUR)
77,90
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
77,90
Each
Chemical Identifiers
CAS579-43-1
IUPAC Name1,2-diphenylethane-1,2-diol
Molecular FormulaC14H14O2
InChI KeyIHPDTPWNFBQHEB-UHFFFAOYNA-N
SMILESOC(C(O)C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystalline and/or lumpy powder
Assay (GC)≥94.0% (sum of isomers)
Melting Point (clear melt)134-140?C
Appearance (Color)White
meso- Hydrobenzoin has been used in the preparation of trans-methyl meso- hydrobenzoin phosphite.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
meso- Hydrobenzoin has been used in the preparation of trans-methyl meso- hydrobenzoin phosphite.

Solubility
Soluble in hot alcohol or chloroform.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible material is strong oxidizing agents.
RUO – Research Use Only

General References:

  1. M. Gary Newton; Bradley S. Campbell. Preparation and crystal structures of trans-methyl meso-hydrobenzoin phosphite and phosphate. J. Am. Chem. Soc. 1974, 96 (25), 7790-7797.
  2. Giovanni Nicolosi; Angela Patti; Mario Piattelli; Claudia Sanfilippo. Desymmetrization of meso-hydrobenzoin via stereoselective enzymatic esterification. Tetrahedron: Asymmetry. 1994, 5 (2), 283-288.