1-Naphthylacetamide, 98%
1-Naphthylacetamide, 98%
1-Naphthylacetamide, 98%
Thermo Scientific Chemicals

1-Naphthylacetamide, 98%

CAS: 86-86-2 | C12H11NO | 185.226 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number B23986.14
also known as B23986-14
Price (EUR)
31,40
Each
Quantity:
25 g
Request bulk or custom format
Price (EUR)
31,40
Each
Chemical Identifiers
CAS86-86-2
IUPAC Name2-(naphthalen-1-yl)acetamide
Molecular FormulaC12H11NO
InChI KeyXFNJVKMNNVCYEK-UHFFFAOYSA-N
SMILESNC(=O)CC1=C2C=CC=CC2=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥97.5%
Appearance (Color)White to pale cream
FormPowder
Melting Point (clear melt)178.0-186.0?C (may be cloudy melt)
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only