2,5-Difluorobenzeneboronic acid, 96%
2,5-Difluorobenzeneboronic acid, 96%
2,5-Difluorobenzeneboronic acid, 96%
2,5-Difluorobenzeneboronic acid, 96%
Thermo Scientific Chemicals

2,5-Difluorobenzeneboronic acid, 96%

CAS: 193353-34-3 | C6H5BF2O2 | 157.911 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number B24113.14
also known as B24113-14
Price (EUR)
194,00
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (EUR)
194,00
Each
Add to cart
Chemical Identifiers
CAS193353-34-3
IUPAC Name(2,5-difluorophenyl)boronic acid
Molecular FormulaC6H5BF2O2
InChI KeyKTOJGSDLJNUAEP-UHFFFAOYSA-N
SMILESOB(O)C1=CC(F)=CC=C1F
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Aqueous acid-base Titration)≥95.0%
FormCrystals or powder or crystalline powder or fused solid
Appearance (Color)White to cream to pale gray
Assay (HPLC)≥95.0%
Proton NMRConforms to structure
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only