Bismuth subgallate hydrate
Bismuth subgallate hydrate
Bismuth subgallate hydrate
Bismuth subgallate hydrate
Thermo Scientific Chemicals

Bismuth subgallate hydrate

CAS: 342406-26-2 | C7H5BiO6 | 394.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number B24187.22
also known as B24187-22
Price (EUR)
36,20
Each
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (EUR)
36,20
Each
Add to cart
Chemical Identifiers
CAS342406-26-2
SpecificationsSpecification SheetSpecification Sheet
Impurity contentNitrate content ≤ 0.2%
Appearance (Color)Yellow
Complexometric Titration48.0-51.0% (dry wt. basis)
Impurity contentLead ≤ 20ppm
Chloride content≤ 200ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only