2-(Bromomethyl)benzonitrile, 98%
2-(Bromomethyl)benzonitrile, 98%
2-(Bromomethyl)benzonitrile, 98%
2-(Bromomethyl)benzonitrile, 98%
Thermo Scientific Chemicals

2-(Bromomethyl)benzonitrile, 98%

CAS: 22115-41-9 | C8H6BrN | 196.05 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number B24278.14
also known as B24278-14
Price (EUR)
117,00
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (EUR)
117,00
Each
Add to cart
Chemical Identifiers
CAS22115-41-9
IUPAC Name2-(bromomethyl)benzonitrile
Molecular FormulaC8H6BrN
InChI KeyQGXNHCXKWFNKCG-UHFFFAOYSA-N
SMILESBrCC1=CC=CC=C1C#N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to dark cream to pale cream
Assay (GC)≥97.5%
Melting Point (clear melt)69.0-76.0?C
FormCrystals or powder or crystalline powder or lumps
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only