1-Amino-4-hydroxyanthraquinone, 96%
1-Amino-4-hydroxyanthraquinone, 96%
1-Amino-4-hydroxyanthraquinone, 96%
Thermo Scientific Chemicals

1-Amino-4-hydroxyanthraquinone, 96%

CAS: 116-85-8 | C14H9NO3 | 239.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number B24388.14
also known as B24388-14
Price (EUR)
123,00
Each
Quantity:
25 g
Request bulk or custom format
Price (EUR)
123,00
Each
Chemical Identifiers
CAS116-85-8
IUPAC Name1-amino-4-hydroxy-9,10-dihydroanthracene-9,10-dione
Molecular FormulaC14H9NO3
InChI KeyAQXYVFBSOOBBQV-UHFFFAOYSA-N
SMILESNC1=CC=C(O)C2=C1C(=O)C1=CC=CC=C1C2=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Appearance (Color)Dark brown to purple
Assay (HPLC)≥95.0%
The use of the inclusion complex of 1-amino-4-hydroxyanthraquinone (AHA) in the internal cavity of β-cyclodextrin confers increased sensitivity, selectivity and detection limit, and allows the determination of 10-70 ng ml-1 beryllium, compared to 60-500 ng ml-1 in the absence of the cyclodextrin.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
The use of the inclusion complex of 1-amino-4-hydroxyanthraquinone (AHA) in the internal cavity of β-cyclodextrin confers increased sensitivity, selectivity and detection limit, and allows the determination of 10-70 ng ml-1 beryllium, compared to 60-500 ng ml-1 in the absence of the cyclodextrin.

Solubility
Soluble in water, ethanol, acetone, benzene.

Notes
Store at room temperature. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. F. Salinas, C. Genestar.; F. Grases. Kinetic-fluorimetric determination of cerium with 1-amino-4-hydroxyanthraquinone.Microchem. J. 1982, 27 (1), 32-36 .
  2. F.Garcia Sanchez.; M.Hernandez Lopez. Spectrofluorimetric determination of beryllium based on the inclusion complex of 1-amino-4-hydroxyanthraquinone with β-cyclodextrin as ligand. Anal. Chim. Acta. 1986, 187 147-155 .