2,6-Dichlorotoluene, 99%
2,6-Dichlorotoluene, 99%
2,6-Dichlorotoluene, 99%
Thermo Scientific Chemicals

2,6-Dichlorotoluene, 99%

CAS: 118-69-4 | C7H6Cl2 | 161.025 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number B24463.14
also known as B24463-14
Price (EUR)
26,70
Each
Quantity:
25 g
Request bulk or custom format
Price (EUR)
26,70
Each
Chemical Identifiers
CAS118-69-4
IUPAC Name1,3-dichloro-2-methylbenzene
Molecular FormulaC7H6Cl2
InChI KeyDMEDNTFWIHCBRK-UHFFFAOYSA-N
SMILESCC1=C(Cl)C=CC=C1Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
Refractive Index1.5495-1.5525 @ 20?C
Assay (GC)≥98.5%
FormLiquid
2,6-Dichlorotoluene is an important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,6-Dichlorotoluene is an important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.

Solubility
Soluble in water.

Notes
Store at room temperature. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. D. Bailey, D. Tirrell.; C. Pinazzi, O. Vogl. Functional Polymers. 4. Polymers of 2,4-Dihydroxy-4'-vinylbenzophenone, New Polymeric Ultraviolet Absorbers. Macromolecules. 1978, 11 (2),312-320 .