2-Amino-5-methylbenzenesulfonic acid, 99%
2-Amino-5-methylbenzenesulfonic acid, 99%
2-Amino-5-methylbenzenesulfonic acid, 99%
2-Amino-5-methylbenzenesulfonic acid, 99%
Thermo Scientific Chemicals

2-Amino-5-methylbenzenesulfonic acid, 99%

CAS: 88-44-8 | C7H9NO3S | 187.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
Catalog number B24560.18
also known as B24560-18
Price (EUR)
32,50
Each
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (EUR)
32,50
Each
Add to cart
Chemical Identifiers
CAS88-44-8
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)≥98.5%
FormPowder
Appearance (Color)Pale cream to cream to brown
Assay (Aqueous acid-base Titration)≥98.5 to ≤101.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only