2-(Bromomethyl)naphthalene, 96%
2-(Bromomethyl)naphthalene, 96%
2-(Bromomethyl)naphthalene, 96%
Thermo Scientific Chemicals

2-(Bromomethyl)naphthalene, 96%

CAS: 939-26-4 | C11H9Br | 221.10 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number B24568.22
also known as B24568-22
Price (EUR)
478,00
Each
Quantity:
100 g
Request bulk or custom format
Price (EUR)
478,00
Each
Chemical Identifiers
CAS939-26-4
IUPAC Name2-(bromomethyl)naphthalene
Molecular FormulaC11H9Br
InChI KeyRUHJZSZTSCSTCC-UHFFFAOYSA-N
SMILESBrCC1=CC=C2C=CC=CC2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Melting Point (clear melt)50.0-59.0?C
Appearance (Color)White to pale cream to pale yellow
Assay (GC)≥95.0%
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Soluble in water (reacts), and chloroform.

Notes
Store in a cool, dry place in tightly closed container. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Ilaria Donelli; Paola Taddei; Philippe F Smet; Dirk Poelman; Vincent A Nierstrasz; Giuliano Freddi. Enzymatic surface modification and functionalization of PET: a water contact angle, FTIR, and fluorescence spectroscopy study. Biotechnology and Bioengineering. 2009, 103 (5), 845-856.
  2. Kuppuswamy Arumugam; James E Bollinger; Mark Fink; James P Donahue. Preparation and isolation of dithiolene thiophosphoryl molecules as stable, protected forms of dithiolene ligands. Inorganic Chemistry. 2007, 46 (8), 3283-3288.
  3. Primary or secondary OH groups can be protected as 2-naphthylmethyl (NAP) ethers, e.g. with NaH in DMF. The NAP ether is stable to dilute aqueous HCl or NaOH, but is cleanly hydrogenolyzed with Pd/C; preferential cleavage in the presence of a benzyl ether can be achieved in high yield: J. Org. Chem., 63, 4172 (1998). NAP protection has been utilized in carbohydrate synthesis where stability to 4% TFA in CHCl3, hot 80% AcOH, SnCl2/AgOTf, or HCl/EtOH were demonstrated, conditions under which the 4-methoxybenzyl (PMB) group is cleaved; both NAP and PMB groups are efficiently cleaved with DDQ: Tetrahedron Lett., 41, 169 (2000).
  4. NAP protection of amino groups and other N functions has also been reported. Cleavage was effected under mild condtions with DDQ: Synlett, 2065 (2003).