2-(Trifluoromethyl)benzaldehyde, 98%
2-(Trifluoromethyl)benzaldehyde, 98%
2-(Trifluoromethyl)benzaldehyde, 98%
Thermo Scientific Chemicals

2-(Trifluoromethyl)benzaldehyde, 98%

CAS: 447-61-0 | C8H5F3O | 174.122 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number B24591.22
also known as B24591-22
Price (EUR)
267,00
Each
Quantity:
100 g
Request bulk or custom format
Price (EUR)
267,00
Each
Chemical Identifiers
CAS447-61-0
IUPAC Name2-(trifluoromethyl)benzaldehyde
Molecular FormulaC8H5F3O
InChI KeyZDVRPKUWYQVVDX-UHFFFAOYSA-N
SMILESFC(F)(F)C1=CC=CC=C1C=O
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.4640-1.4680 @ 20°C
Appearance (Color)Clear colorless to pale yellow
FormLiquid
Assay (GC)≥97.5%
Identification (FTIR)Conforms
2-(Trifluoromethyl)benzaldehyde has been used in the preparation of ω-(dimethylamino)-alkylethyl-4(-2-(trifluorormethyl)phenyl)-2,6-dimethyl-1,4-dihydropyridine-3,5-dicarboxylates, methyl (E)-2-[2-[(E)-2-(2-trifluoromethylphenyl)vinyl]-phenyl]-3-methoxyacrylate via Wittig-Horner reaction. It is a reagent used as electrophilic component in a wide array of reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(Trifluoromethyl)benzaldehyde has been used in the preparation of ω-(dimethylamino)-alkylethyl-4(-2-(trifluorormethyl)phenyl)-2,6-dimethyl-1,4-dihydropyridine-3,5-dicarboxylates, methyl (E)-2-[2-[(E)-2-(2-trifluoromethylphenyl)vinyl]-phenyl]-3-methoxyacrylate via Wittig-Horner reaction. It is a reagent used as electrophilic component in a wide array of reactions.

Solubility
Soluble in most organic solvents.

Notes
Air sensitive. Store under dry inert gas. Store away from oxidizing agents, air.
RUO – Research Use Only

General References:

  1. N Baindur; A Rutledge; D J Triggle. A homologous series of permanently charged 1,4-dihydropyridines: novel probes designed to localize drug binding sites on ion channels. Journal of Medicinal Chemistry. 1993, 36 (23), 3743-3745.
  2. J Alzeer, J Chollet; I Heinze-Krauss; C Hubschwerlen; H Matile; R G Ridley. Phenyl beta-methoxyacrylates: a new antimalarial pharmacophore. Journal of Medicinal Chemistry. 2000, 43 (4), 560-568.