4-Pyridineacetamide, 98%
4-Pyridineacetamide, 98%
4-Pyridineacetamide, 98%
Thermo Scientific Chemicals

4-Pyridineacetamide, 98%

CAS: 39640-62-5 | C7H8N2O | 136.154 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number B24734.03
also known as B24734-03
Price (EUR)
19,20
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
19,20
Each
Chemical Identifiers
CAS39640-62-5
IUPAC Name2-(pyridin-4-yl)acetamide
Molecular FormulaC7H8N2O
InChI KeyYEALAJQPOVKTOH-UHFFFAOYSA-N
SMILESNC(=O)CC1=CC=NC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormCrystals or powder or crystalline powder
Assay (GC)≥97.5%
Melting Point (clear melt)141.5-147.5?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only