1-Adamantaneacetonitrile, 97%
1-Adamantaneacetonitrile, 97%
1-Adamantaneacetonitrile, 97%
1-Adamantaneacetonitrile, 97%
Thermo Scientific Chemicals

1-Adamantaneacetonitrile, 97%

CAS: 16269-13-9 | C12H17N | 175.28 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number B24770.03
also known as B24770-03
Price (EUR)
78,10
Each
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (EUR)
78,10
Each
Add to cart
Chemical Identifiers
CAS16269-13-9
IUPAC Name2-(adamantan-1-yl)acetonitrile
Molecular FormulaC12H17N
InChI KeyDXQVPXCZIRQITG-UHFFFAOYSA-N
SMILESN#CCC12CC3CC(CC(C3)C1)C2
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Melting Point (clear melt)71.5-77.5?C
Appearance (Color)White
Assay (GC)≥96.0%
1-Adamantaneacetonitrile is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Adamantaneacetonitrile is used as pharmaceutical intermediate.

Solubility
Sparingly soluble in water.

Notes
Store in a cool, dry place in tightly closed container. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Richard O. AngusJr.; Richard P. Johnson. Small-ring cyclic cumulenes: the structure and energetics of cyclic butatrienes and the synthesis of 1,2,3-cyclononatriene. J. Org. Chem. 1984, 49 (16), 2880-2883.
  2. Mary J. Guttieri; Wilhelm F. Maier. Selective cleavage of carbon-nitrogen bonds with platinum. J. Org. Chem. 1984, 49 (16), 2875-2880.