4-Bromo-2-chlorophenol, 98%
4-Bromo-2-chlorophenol, 98%
4-Bromo-2-chlorophenol, 98%
4-Bromo-2-chlorophenol, 98%
Thermo Scientific Chemicals

4-Bromo-2-chlorophenol, 98%

CAS: 3964-56-5 | C6H4BrClO | 207.45 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
Catalog number B24803.09
also known as B24803-09
Price (EUR)
25,60
Each
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (EUR)
25,60
Each
Add to cart
Chemical Identifiers
CAS3964-56-5
IUPAC Name4-bromo-2-chlorophenol
Molecular FormulaC6H4BrClO
InChI KeyVIBJPUXLAKVICD-UHFFFAOYSA-N
SMILESOC1=CC=C(Br)C=C1Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormPowder or lumps or fused solid
Assay (GC)≥97.5%
Melting Point (clear melt)46.0-53.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only