Methyl 2,6-dichlorobenzoate, 97%
Methyl 2,6-dichlorobenzoate, 97%
Methyl 2,6-dichlorobenzoate, 97%
Thermo Scientific™

Methyl 2,6-dichlorobenzoate, 97%

CAS: 14920-87-7 | C8H6Cl2O2 | 205.03 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number B24912.06
also known as B24912-06
Price (EUR)
97,60
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
97,60
Each
Chemical Identifiers
CAS14920-87-7
IUPAC Namemethyl 2,6-dichlorobenzoate
Molecular FormulaC8H6Cl2O2
InChI KeyHEYGSGDIWJDORA-UHFFFAOYSA-N
SMILESCOC(=O)C1=C(Cl)C=CC=C1Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to very pale yellow
FormLow melting solid (clear liquid as melt)
GC/MS AnalysisConsistent with Molecular mass : 205
Refractive Index1.5255-1.5305 (as melt)
Assay (GC)>96.0%