Coniferyl alcohol, 98%
Coniferyl alcohol, 98%
Coniferyl alcohol, 98%
Thermo Scientific Chemicals

Coniferyl alcohol, 98%

CAS: 458-35-5 | C10H12O3 | 180.20 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
0.25 g
1 g
5 g
Catalog number B24949.01
also known as B24949-01
Price (EUR)
100,65
Online exclusive
112,00
Save 11,35 (10%)
Each
Quantity:
0.25 g
Request bulk or custom format
Price (EUR)
100,65
Online exclusive
112,00
Save 11,35 (10%)
Each
Chemical Identifiers
CAS458-35-5
IUPAC Name4-[(1E)-3-hydroxyprop-1-en-1-yl]-2-methoxyphenol
Molecular FormulaC10H12O3
InChI KeyJMFRWRFFLBVWSI-NSCUHMNNSA-N
SMILESCOC1=CC(C=CCO)=CC=C1O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥97.5%
Identification (FTIR)Conforms
Appearance (Color)White to cream to yellow or pale orange
FormCrystals or powder or crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only