Chloroacetaldehyde dimethyl acetal, 97%
Chloroacetaldehyde dimethyl acetal, 97%
Chloroacetaldehyde dimethyl acetal, 97%
Thermo Scientific Chemicals

Chloroacetaldehyde dimethyl acetal, 97%

CAS: 97-97-2 | C4H9ClO2 | 124.56 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number B25049.22
also known as B25049-22
Price (EUR)
27,60
Each
Quantity:
100 g
Request bulk or custom format
Price (EUR)
27,60
Each
Chemical Identifiers
CAS97-97-2
IUPAC Name2-chloro-1,1-dimethoxyethane
Molecular FormulaC4H9ClO2
InChI KeyCRZJPEIBPQWDGJ-UHFFFAOYSA-N
SMILESCOC(CCl)OC
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Appearance (Color)Clear colorless to yellow
Assay (GC)≥97.5%
Identification (FTIR)Conforms
Refractive Index1.4130-1.4170 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only