1,3-Di-o-tolylguanidine, 99%
1,3-Di-o-tolylguanidine, 99%
1,3-Di-o-tolylguanidine, 99%
Thermo Scientific Chemicals

1,3-Di-o-tolylguanidine, 99%

CAS: 97-39-2 | C15H17N3 | 239.322 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number B25127.36
also known as B25127-36
Price (EUR)
196,00
Each
Quantity:
500 g
Request bulk or custom format
Price (EUR)
196,00
Each
Chemical Identifiers
CAS97-39-2
IUPAC NameN,N''-bis(2-methylphenyl)guanidine
Molecular FormulaC15H17N3
InChI KeyOPNUROKCUBTKLF-UHFFFAOYSA-N
SMILESCC1=CC=CC=C1NC(N)=NC1=CC=CC=C1C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream
Assay (Non-aqueous acid-base Titration)≥98.5 to ≤101.5%
Melting Point (clear melt)171.0-177.0?C
FormPowder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only