1-(2-Phenylethyl)piperazine, 98%
1-(2-Phenylethyl)piperazine, 98%
1-(2-Phenylethyl)piperazine, 98%
Thermo Scientific Chemicals

1-(2-Phenylethyl)piperazine, 98%

CAS: 5321-49-3 | C12H18N2 | 190.29 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number B25158.09
also known as B25158-09
Price (EUR)
42,50
Each
Quantity:
10 g
Request bulk or custom format
Price (EUR)
42,50
Each
Chemical Identifiers
CAS5321-49-3
IUPAC Name1-(2-phenylethyl)piperazine
Molecular FormulaC12H18N2
InChI KeyLKUAPSRIYZLAAO-UHFFFAOYSA-N
SMILESC(CC1=CC=CC=C1)N1CCNCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.5410-1.5450 @ 20°C (non-U.S. specification)
FormLiquid
Appearance (Color)Clear colorless to yellow
Assay (GC)≥97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only