Methyl 4-iodo-3-methylbenzoate is used in the synthesis of novel colchicine-derived nitrate esters as immunosuppressant agents.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Methyl 4-iodo-3-methylbenzoate is used in the synthesis of novel colchicine-derived nitrate esters as immunosuppressant agents.
Solubility
Slightly soluble in methanol.
Notes
Light sensitive. Store away from oxidizing agents, light.
RUO – Research Use Only
General References:
- Laurie F. Mottram; Ewa Maddox; Markus Schwab; Florent Beaufils and Blake R. Peterson. A Concise Synthesis of the Pennsylvania Green Fluorophore and Labeling of Intracellular Targets with O6-Benzylguanine Derivatives. Org. Lett. 2007, 9 (19), 3741-3744.
- Xijun Piao; Ying Zou; Junchen Wu; Chunyan Li and Tao Yi. Multiresponsive Switchable Diarylethene and Its Application in Bioimaging. Org. Lett. 2007, 11 (17), 3818-3821.