2-Amino-5-nitrobenzonitrile, 95%
2-Amino-5-nitrobenzonitrile, 95%
2-Amino-5-nitrobenzonitrile, 95%
2-Amino-5-nitrobenzonitrile, 95%
Thermo Scientific Chemicals

2-Amino-5-nitrobenzonitrile, 95%

CAS: 17420-30-3 | C7H5N3O2 | 163.136 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number B25416.22
also known as B25416-22
Price (EUR)
54,80
Each
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (EUR)
54,80
Each
Add to cart
Chemical Identifiers
CAS17420-30-3
IUPAC Name2-amino-5-nitrobenzonitrile
Molecular FormulaC7H5N3O2
InChI KeyMGCGMYPNXAFGFA-UHFFFAOYSA-N
SMILESNC1=CC=C(C=C1C#N)[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Melting Point208.0-214.0?C
Appearance (Color)Yellow to orange
Assay (GC)≥94.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only