1-Iodo-4-isopropylbenzene, 97%
1-Iodo-4-isopropylbenzene, 97%
1-Iodo-4-isopropylbenzene, 97%
Thermo Scientific Chemicals

1-Iodo-4-isopropylbenzene, 97%

CAS: 17356-09-1 | C9H11I | 246.091 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number B25476.06
also known as B25476-06
Price (EUR)
60,10
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
60,10
Each
Chemical Identifiers
CAS17356-09-1
IUPAC Name1-iodo-4-(propan-2-yl)benzene
Molecular FormulaC9H11I
InChI KeyPQJOSEVTIKYWLH-UHFFFAOYSA-N
SMILESCC(C)C1=CC=C(I)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
Refractive Index1.5755-1.5805 @ 20?C
Assay (GC)≥96.0%
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only