2,4-Difluoro-3-methoxybenzoic acid, 98+%
2,4-Difluoro-3-methoxybenzoic acid, 98+%
2,4-Difluoro-3-methoxybenzoic acid, 98+%
2,4-Difluoro-3-methoxybenzoic acid, 98+%
Thermo Scientific Chemicals

2,4-Difluoro-3-methoxybenzoic acid, 98+%

CAS: 178974-97-5 | C8H6F2O3 | 188.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number B25597.14
also known as B25597-14
Price (EUR)
239,00
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (EUR)
239,00
Each
Add to cart
Chemical Identifiers
CAS178974-97-5
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormCrystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)≥98.0 to ≤102.0%
Assay (HPLC)≥98.0%
Melting Point (clear melt)191.0-197.0?C
2,4-Difluoro-3-methoxybenzoic acid is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4-Difluoro-3-methoxybenzoic acid is used as a pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
RUO – Research Use Only

General References:

  1. Liu; Kevin K.-C.; Sakya; Subas M.; O'Donnell; Christopher J.; Li, Jin. Synthetic Approaches to the 2007 New Drugs. Mini Reviews in Medicinal Chemistry. 2008, 8 (14), 1526-1548.
  2. Graul, A.; Rabasseda, X.; Castaner, J. T-3811ME. Drugs Fut. 1999, 24 (12), 1324.