(+/-)-1,2,3,4-Tetrahydro-1-naphthylamine, 97%
(+/-)-1,2,3,4-Tetrahydro-1-naphthylamine, 97%
(+/-)-1,2,3,4-Tetrahydro-1-naphthylamine, 97%
(+/-)-1,2,3,4-Tetrahydro-1-naphthylamine, 97%
Thermo Scientific Chemicals

(+/-)-1,2,3,4-Tetrahydro-1-naphthylamine, 97%

CAS: 2217-40-5 | C10H14ClN | 183.68 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number B25673.14
also known as B25673-14
Price (EUR)
139,00
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (EUR)
139,00
Each
Add to cart
Chemical Identifiers
CAS2217-40-5
IUPAC Name1,2,3,4-tetrahydronaphthalen-1-aminium chloride
Molecular FormulaC10H14ClN
InChI KeyDETWFIUAXSWCIK-UHFFFAOYNA-N
SMILES[Cl-].[NH3+]C1CCCC2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow to brown
Assay (GC)≥96.0%
Refractive Index1.5615-1.5665 @ 20?C
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only