5-Bromo-2-(trifluoromethyl)pyridine, 97%
5-Bromo-2-(trifluoromethyl)pyridine, 97%
5-Bromo-2-(trifluoromethyl)pyridine, 97%
Thermo Scientific Chemicals

5-Bromo-2-(trifluoromethyl)pyridine, 97%

CAS: 436799-32-5 | C6H3BrF3N | 225.996 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number B25731.03
also known as B25731-03
Price (EUR)
91,30
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
91,30
Each
Chemical Identifiers
CAS436799-32-5
IUPAC Name5-bromo-2-(trifluoromethyl)pyridine
Molecular FormulaC6H3BrF3N
InChI KeyRPFAUCIXZGMCFN-UHFFFAOYSA-N
SMILESFC(F)(F)C1=CC=C(Br)C=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormPowder or crystalline powder
Assay (GC)≥96.0%
Identification (FTIR)Conforms
Melting Point (clear melt)39-44?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
 
RUO – Research Use Only