Perform accurate pH calibrations with color-coded, twin neck pH Buffer Bottles.
Have Questions?
Change view
Catalog Number
pH
BS04500ML
also known as BS04/500ML
4.01
BS07500ML
also known as BS07/500ML
7.00
BS10500ML
also known as BS10/500ML
10.01
3 Options
Catalog number BS04500ML
also known as BS04/500ML
Price (EUR)
10,53
キャンペーン価格
11,70
Save 1,17 (10%)
Each
pH:
4.01
Price (EUR)
10,53
キャンペーン価格
11,70
Save 1,17 (10%)
Each
Perform accurate pH calibrations with color-coded Thermo Scientific™ pH Buffer Bottles. The bottles have a twin neck and the pH 4.01, 7.00 and 10.01 buffer bottles are color coded for ease of use.
Buffer Color Codes:
Pink = pH 4.01
Yellow = pH 7.00
Blue = pH 10.01
Available in 500 mL bottles
Specifications
Boiling Point100°C
ColorLight Red
DescriptionBuffer
FormulationWater, Potassium Hydroxide, Amaranth Red No. 27, Potassium Hydrogen Phthalate