Product Image
Thermo Scientific™

Thermo Scientific™ pH Buffers, Twin Neck

Perform accurate pH calibrations with color-coded, twin neck pH Buffer Bottles.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberpH
BS10500ML
also known as BS10/500ML
10.01
BS04500ML
also known as BS04/500ML
4.01
BS07500ML
also known as BS07/500ML
7.00
Catalog number BS10500ML
also known as BS10/500ML
Price (EUR)
10,53
Online exclusive
11,70
Save 1,17 (10%)
Each
pH:
10.01
Price (EUR)
10,53
Online exclusive
11,70
Save 1,17 (10%)
Each
Perform accurate pH calibrations with color-coded Thermo Scientific™ pH Buffer Bottles. The bottles have a twin neck and the pH 4.01, 7.00 and 10.01 buffer bottles are color coded for ease of use.

Buffer Color Codes:

  • Pink = pH 4.01
  • Yellow = pH 7.00
  • Blue = pH 10.01

Available in 500 mL bottles

Specifications
Boiling Point100°C
ColorLight Blue
DescriptionBuffer
FormulationWater, Sodium Bicarbonate, Methylparaben, FD & C Blue #1, Sodium Carbonate
Solubility InformationSoluble
For Use With (Application)pH calibration
Physical FormLiquid
Quantity500 mL
Solution TypeBuffer
pH10.01
Unit SizeEach