Product Image
Thermo Scientific™
Eutech™ Ion Selective Combination Electrode, Epoxy-Body
Use Eutech™ Ion Selective Combination Electrode for measurement of specific ions in aqueous solutions.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberElectrode
CAL0301BCalcium
BRO0301BBromide
FO0301BFluoride
IO0301BIodide
NH40301BAmmonium
Catalog number CAL0301B
Price (EUR)
-
Electrode:
Calcium
Eutech™ offers a wide selection of Ion Selective Electrodes (ISE) and meters for the measurement of ammonia, bromide, cadmium, calcium, carbon dioxide, carbonate, chloride, copper, cyanide, fluoride, iodide, lead, nitrate, potassium, silver, sulfide and sodium.ISE responds to ionic activity or concentration in a liquid because of the characteristics of the electrode’s sensing membrane. The ISE develops an electrical potential which is proportional to the activity or concentration of the ion for which the membrane is selective.
Features:
  • Epoxy-body
  • For Use with: IS meters
Specifications
ElectrodeCalcium
For Use With (Equipment)Eutech Ion Meters
MaterialEpoxy
TypeCombination ISE
Unit SizeEach