InstaFill™ Peristaltic Pump
InstaFill™ Peristaltic Pump
InstaFill™ Peristaltic Pump
InstaFill™ Peristaltic Pump
Thermo Scientific™
InstaFill™ Peristaltic Pump
The Thermo Scientific™ InstaFill™ Peristaltic Pump provides automated volumetric media dispensing for precise, traceable sample preparation workflows. Dispensed volumes can be calibrated against a gravimetric equivalent where required.
Have Questions?
Catalog number DB7001IN
Price (EUR)
-
The Thermo Scientific™ InstaFill™ Peristaltic Pump provides controlled hydration of InstaPrep™ 10x media bags, supporting reproducible media preparation as part of the InstaFlux™ workflow. By standardizing the hydration step, InstaFill™ ensures consistent media concentration prior to dilution, supporting downstream workflows aligned with ISO microbiological standards. The InstaFill™ Peristaltic Pump will also function as a standard peristaltic pump suitable for routine laboratory applications.
Key Features and Product Benefits
  • Controlled hydration for reproducible media preparation, including calibration capability
  • Suitable for dispensing media into large volume samples (once suitably calibrated)
  • Supports standardized workflows aligned with ISO methods
  • High-flow pump for rapid media preparation
  • Integrates seamlessly into the InstaFlux™ system
  • Barcode integration and traceability
Sample Preparation
Ensures consistent preparation of enrichment media prior to dilution, supporting reliable performance in ISO-aligned microbiological workflows.
Part of a Suite
Works with InstaPrep™ media, InstaDose™ Gravimetric Diluter and Peristaltic Pump, and InstaFill™ tubing and accessories.
For Testing of Food and Environmental Samples Only.
Specifications
Dimensions (L x W x H)253 x 160 x 250 mm
Electrical RequirementsAC 85 to 264 Volts / 47 to 63 Hz
For Use With (Application)InstaFlux™ Workflow
For Use With (Equipment)InstaFill™ tubing for InstaPrep™ / DryBags™ hydration
IncludesInstaFill™ Peristaltic Pump, Power cord, Power supply
Wattage120 W
Weight (Metric)3.1 kg
Voltage80/240 V
Unit SizeEach