TDS 442 Standard Solution, Eutech™
Thermo Scientific™

TDS 442 Standard Solution, Eutech™

For TDS calibration and standards, choose from our available Eutech™'s TDS Standards, Calibrations, and Solutions
Have Questions?
Change viewbuttonViewtableView
Catalog NumberChemical Name or MaterialConcentration
EC442300BT442 Standard Solution
EC4421000BT442 Standard Solution
EC4423000BT442 Calibration Solution
EC44250BT442 Calibration Solution
Catalog number EC442300BT
Price (EUR)
14,46
線上優惠
16,07
Save 1,61 (10%)
Each
Chemical Name or Material:
442 Standard Solution
Price (EUR)
14,46
線上優惠
16,07
Save 1,61 (10%)
Each

Eutech™ TDS Standard Solutions are used as standards for TDS meters. Get quick and accurate conductivity TDS measurements using Eutech™ TDS meters in the field, laboratories, and schools.

Features:

  • Buffers and calibration solutions packed in 480 ml durable plastic bottles – economical and convenient for laboratory use.
Specifications
Boiling Point∼100°C
Chemical Name or Material442 Standard Solution
ColorColorless
PackagingPlastic Bottle
Physical FormLiquid
Quantity480 mL
Solution TypeStandard Solution
TypeTDS Standard
Industry TypeLaboratory Use
pH7
Unit SizeEach