Product Image
Thermo Scientific™

Eutech™ EC620131 pH electrode

Eutech™ offers a wide range of pH meters and electrodes that measure pH values in various educational, laboratory and industrial applications.
Have Questions?
Catalog NumberConnector Types
EC620131BNC
Catalog number EC620131
Price (EUR)
170,10
온라인 행사
189,00
Save 18,90 (10%)
Each
Connector Types:
BNC
Price (EUR)
170,10
온라인 행사
189,00
Save 18,90 (10%)
Each

The measure of a liquid's acidity and alkalinity, pH is one of the most common parameter measured in a wide variety of industries ranging from water and wastewater treatment, chemical production, agriculture research and production, environmental monitoring, chemical and life sciences research, biochemical and pharmaceutical research, electronics production to food processing and other industrial applications.

Eutech™ offers a wide range of pH meters and electrodes that measure pH values in various educational, laboratory and industrial applications.

Features:

  • Buffers and standard solutions packed in 480 ml durable plastic bottles – economical and convenient for laboratory use.
Specifications
DescriptionOpen pore glass-body pH electrode for general testing and high viscosity solutions where frequent cleaning of reference is required
For Use With (Application)pH measurement
For Use With (Equipment)All pH meters with BNC input connector
Temperature (Metric) Range0°C to 80°C
Connector TypesBNC
MaterialGlass Body
Temperature (Metric) Operating0°C to 80°C
TypepH Electrode
pH Range0 to 14
Unit SizeEach