Product Image
Thermo Scientific™
Eutech™ Epoxy-body Conductivity Electrode for CyberScan CON 1500
Get quick and accurate conductivity measurements in the field, laboratories, and schools.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCell Constant
EC620160K = 1.0
EC620162K = 10.0
Catalog number EC620160
Price (EUR)
260,10
온라인 행사
289,00
Save 28,90 (10%)
Each
Cell Constant:
K = 1.0
Price (EUR)
260,10
온라인 행사
289,00
Save 28,90 (10%)
Each
Eutech™ Conductivity Electrodes for CyberScan CON 1500 are used in conductivity meters in various applications. Get quick and accurate conductivity measurements in the field, laboratories, and schools.
Features
  • Parameter: Conductivity
  • Dual Pin Connector
Specifications
DescriptionEpoxy-body 2-ring Conductivity Electrode, cell constant K=1.0, dual pin connector, 1m cable (for CON 1500)
For Use With (Equipment)CyberScan CON 1500
Length (Metric) Cable1 m
Cell ConstantK = 1.0
Connector TypesDual Pin
MaterialEpoxy-body 2-ring
TypeConductivity Electrode
Unit SizeEach