Product Image
Thermo Scientific™

Eutech™ ECFG7370101B pH electrode

Eutech™ offers a wide range of pH meters and electrodes that measure pH values in various educational, laboratory and industrial applications.
Have Questions?
Catalog NumberConnector Types
ECFG7370101BBNC
Catalog number ECFG7370101B
Price (EUR)
243,00
キャンペーン価格
270,00
Save 27,00 (10%)
Each
Connector Types:
BNC
Price (EUR)
243,00
キャンペーン価格
270,00
Save 27,00 (10%)
Each
The measure of a liquid acidity and alkalinity, pH is one of the most common parameter measured in a wide variety of industries ranging from water and wastewater treatment, chemical production, agriculture research and production, environmental monitoring, chemical and life sciences research, biochemical and pharmaceutical research, electronics production to food processing and other industrial applications. Eutech™ offers a wide range of pH meters and electrodes that measure pH values in various educational, laboratory and industrial applications. This clog free double-junction glass pH electrode is ideal for samples containing TRIS buffers, sulfides and more. Easy to operate fill hole sleeve mechanism is used for general pH measurements. Available with 1 m cable.

Specifications

  • Parameter: pH
  • Range: 0 to 13 pH
  • Temperature Range: 0 to 100 °C
  • Liquid Junction Type: Annular ceramic
  • Internal Reference Type: Ag/AgCl
  • Sealed/Refillable: Refillable
  • Reference Junction: Double
  • Refilling Reference Electrolyte: ECRE002
  • Dimensions (Shaft): 110 x 12mm
  • Cable Length: 1m
  • Connector: BNC
  • Used With: All pHmeters with BNC input connector
  • Specifications
    Connector TypesBNC
    JunctionDouble
    Temperature (Metric) Operating0°C to 100°C
    TypepH Electrode
    pH Range0 to 13
    Unit SizeEach