Product Image
Thermo Scientific™
Eutech™ pH electrode ECFGHGK3F01B
Eutech™ offers a wide range of pH meters and electrodes that measure pH values in various educational, laboratory and industrial applications.
Have Questions?
Catalog NumberLength (Metric)
ECFGHGK3F01B110 mm
Catalog number ECFGHGK3F01B
Price (EUR)
177,30
온라인 행사
197,00
Save 19,70 (10%)
Each
Length (Metric):
110 mm
Price (EUR)
177,30
온라인 행사
197,00
Save 19,70 (10%)
Each
Eutech™ offers a wide range of pH meters and electrodes that measure pH values in various educational, laboratory and industrial applications. Eutech™ pH electrode ECFGHGK3F01B is a reinforced glass-body refillable pH Electrode with a spear tip for direct soil pH measurement.
Features:
  • Glass body
  • Refillable
Specifications
DescriptionReinforced Glass-body Refillable pH Electrode, spear tip for direct soil measurement
Diameter (Metric) Body12 mm
For Use With (Application)pH
For Use With (Equipment)All pH meters with BNC input connector
Length (Metric)110 mm
Length (Metric) Cable1 m
Tip TypeSpear
Connector TypesBNC
MaterialGlass Body
TypepH Electrode
Unit SizeEach