Eutech™ Ion 2700 Meter
Eutech™ Ion 2700 Meter
Eutech™ Ion 2700 Meter
Thermo Scientific™

Eutech™ Ion 2700 Meter

Quick and easy measurement with this ion-selective meter.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberProduct Type
ECION270042GSMultiparameter Meter Kit
ECION270040SMultiparameter Meter
Catalog number ECION270042GS
Price (EUR)
1.096,86
Online exclusive
1.218,73
Save 121,87 (10%)
Each
Product Type:
Multiparameter Meter Kit
Price (EUR)
1.096,86
Online exclusive
1.218,73
Save 121,87 (10%)
Each
Intuitive, self-diagnostic and flexible with advanced set-up options for user-customization, the Eutech ION 2700 series comes with a large, one-glance-sees-all screen. View pH, ION or Redox reading together with temperature, electrode status, calibration points, date and time all at once. All configurations include 30X427301 RS232 cable, 100/240 VAC power adapter and an integral electrode holder.

Features:

  • Up to 6-point push button calibration with auto-buffer recognition
  • Quick, easy electrode diagnosis with pH slope and offset display
  • Non-volatile memory holds up to 500 data points – time and date-stamped for GLP compliance
  • Bi-directional RS232 for easy data transfer to computer
  • Cal-due alarm – no more out-dated calibrations!
  • Auto-logging function for convenient continuous monitoring
  • Limit alarm alerts when reading falls out of range
  • Password protection for setup and calibration
  • Buffer Sets: USA, NIST, DIN, User 1, User 2, Custom

Applications:

  • Educational: Useful for most laboratory, ecological studies and other applications.
  • Laboratory: Environmental studies, chemical labs, titrations and quality assurance testing, where GLP data-management is required. Use in all types of food processing
Specifications
Accuracy (ISE)0.5% full scale (monovalent ion), 1% full scale (divalent ion)
Accuracy (ORP)±0.2 mV
Accuracy (pH)±0.002 + 1 LSD
Calibration PointsUp to 6 preset or 5 custom
Calibration Points (ISE)2 to 8
Calibration Points (pH)Up to 6 preset or 5 custom
DescriptionEutech ION 2700 Meter With pH Electrode (ECFG7370101B), ATC Probe (PH5TEMB01P) & pH Electrode Refill Solution, 60 mL
IncludesIntegral Electrode Holder & 100/240 VAC Adapter and RS232 cable
OutputsRS232
Power Supply9 V DC adapter, 1.3 A (100/240 VAC, SMPS)
Product TypeMultiparameter Meter Kit
Range (Concentration)0.001 to 19999 ppm (±2000 mV)
Range (DO)±2000.0 mV
Range (ORP)±2000.0 mV
Range (Relative mV)±2000.0 mV
Resolution (ISE)2 or 3 digits
Resolution (ORP)0.1 mV
Resolution (pH)0.1/0.01/0.001
Temperature Accuracy±0.3°C, ±0.5°F
Temperature CompensationATC / MTC (0 to 100 °C) (pH only)
Temperature Range (English)32.0°F to 212.0°F
Temperature Resolution0.1 °C / 0.1 °F
Measurement ModespH, Ion, Redox
Range (mV)±2000.0 mV
Range (pH)-2 to 20
Temperature Range (Metric)0.0°C to 100.0°C
Unit SizeEach