Product Image
Thermo Scientific™
Eutech™ ORP Solutions
Eutech™ offers ORP Standard Solutions for use with ORP Meters.
Have Questions?
Catalog number ECORPPRE
Price (EUR)
32,14
온라인 행사
35,71
Save 3,57 (10%)
Each
Price (EUR)
32,14
온라인 행사
35,71
Save 3,57 (10%)
Each
ORP meters can measure both pH and ORP values in various educational, laboratory and industrial applications. Major areas of usage include the treatment of industrial wastes, study of biological systems, oxidation of cyanide, bleaching of pulp, manufacture of bleach and reduction of chromate wastes. The measurement of ORP is also useful in pool water treatment as an indication of sanitation in relation to free chlorine parameter. Eutech™ offers a range of ORP Solutions for use with our ORP Meters.
Features:
  • Buffers and calibration solutions packed in 480 ml durable plastic bottles – economical and convenient for laboratory use.
Specifications
DescriptionPre-treatment solution (475 mV)
For Use With (Equipment)ORP Meter
Unit SizeEach