Eutech™ pH 5+ pH meter
Eutech™ pH 5+ pH meter
Eutech™ pH 5+ pH meter
Thermo Scientific™

Eutech™ pH 5+ pH meter

Eutech™ offers a wide range of pH meters and electrodes that measure pH values in various educational, laboratory and industrial applications.
Have Questions?
Catalog NumberCalibration Points
ECPH502PLUSK2 to 5
Catalog number ECPH502PLUSK
Price (EUR)
382,50
Online exclusive
425,00
Save 42,50 (10%)
Each
Calibration Points:
2 to 5
Price (EUR)
382,50
Online exclusive
425,00
Save 42,50 (10%)
Each
Eutech™ offers a wide range of pH meters and electrodes that measure pH values in various educational, laboratory and industrial applications. Eutech™ pH 5+ pH Meter with single Junction pH electrode is an ideal choice for routine applications in laboratories, productions plants and schools.

Features:

  • multi-Point Calibration (up to 5 points) with Auto-Buffer Recognition - choice of USA, NIST or pure water buffer standards with ±0.01 pH accuracy
  • Automatic Temperature Compensation (ATC) for the highest accuracy under fluctuating temperatures
  • Hold Functionmomentarily freezes reading for easy viewing and recording
  • Auto-Power Off saves battery power after non-use
  • Self-Diagnostic withmessage codes for easy troubleshooting
  • Large Custom LCD provides optimum viewing even at a distance
  • Protective Rubber Boot shieldsmeter from accidental knocks and features a sturdy built-in stand for easy bench-top operation
  • Rugged Carrying Case (Optional) provides handy on-sitemeter calibration andmeasurement

Specifications

  • pH Range: 0.00 to 14.00 pH
  • Temperature Range: 0 to 100.0 °C
  • Temperature Compensation: Automatic /manual (0 to 100 °C)
  • No. of Calibration Points: 2 to 5 points
  • pH Buffer Options:1.68, 4.01, 7.00, 10.01, 12.45 (USA), 1.68, 4.01, 6.86, 9.18, 12.45 (NIST), 4.10, 6.97 (Pb)
  • Special Functions: Auto Power-Off after 17minutes;
  • Hold & Self-diagnosticmessages
  • Power: 4 'AAA' x 1.4V batteries
Specifications
DescriptionEutech pH 5+ pH Meter with Single Junction pH Electrode ECFC7252101B, ATC Electrode ECPHTEM01P & pH Carrying Kit Set (Includes 1 x pH 4.01, pH 7.00 Buffer Solutions, Storage Solution & 1 x Deionised (Rinse) Water (60 ml Each)
Calibration Points2 to 5
Display TypeLCD
Temperature Range (Metric)0°C to 100°C
Unit SizeEach