1,1-Cyclopropanedicarboxylic acid monomethyl ester, 97%
1,1-Cyclopropanedicarboxylic acid monomethyl ester, 97%
1,1-Cyclopropanedicarboxylic acid monomethyl ester, 97%
Thermo Scientific Chemicals

1,1-Cyclopropanedicarboxylic acid monomethyl ester, 97%

CAS: 113020-21-6 | C6H8O4 | 144.126 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H25828.03
also known as H25828-03
Price (EUR)
27,30
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
27,30
Each
Chemical Identifiers
CAS113020-21-6
IUPAC Name1-(methoxycarbonyl)cyclopropane-1-carboxylic acid
Molecular FormulaC6H8O4
InChI KeyIZYOHLOUZVEIOS-UHFFFAOYSA-N
SMILESCOC(=O)C1(CC1)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Assay (GC)≥96.0%
FormCrystals or powder or crystalline powder
1,1-Cyclopropanedicarboxylic acid monomethyl ester is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,1-Cyclopropanedicarboxylic acid monomethyl ester is used as a pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Store in a cool, dry place in tightly closed container. Incompatible with oxidizing agents.
RUO – Research Use Only