3-Fluoro-4-methoxybenzyl bromide, 98%
3-Fluoro-4-methoxybenzyl bromide, 98%
3-Fluoro-4-methoxybenzyl bromide, 98%
Thermo Scientific Chemicals

3-Fluoro-4-methoxybenzyl bromide, 98%

CAS: 331-61-3 | C8H8BrFO | 219.05 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H26116.03
also known as H26116-03
Price (EUR)
127,00
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
127,00
Each
Chemical Identifiers
CAS331-61-3
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale cream to cream
FormCrystals or powder or crystalline powder
Assay (GC)≥97.5%
Melting Point (clear melt)41.0-47.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only